Gene Transcripts

Gene Symbol
Itga8
Entrez Gene ID
364786
Gene Name
integrin, alpha 8
Gene Aliases
RGD1564327
Chromosome Location
Chr. 17: 79321896 - 79676499 on Build Rnor_6.0
UniGene ID
Rn.69726
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001173972.1 NP_001167443.1 13 1524
GenBank mRNA BC168655.1 - 13 1524

View Full Product Details