Gene Transcripts

Gene Symbol
Aqp4
Entrez Gene ID
25293
Gene Name
aquaporin 4
Gene Aliases
AQP-4, MIWC2, Miwc
Chromosome Location
Chr. 18: 6766009 - 6782757 on Build Rnor_6.0
UniGene ID
Rn.90091
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142366.2 NP_001135838.1 2 291
NM_001270558.2 NP_001257487.1 2 283
NM_001270559.2 NP_001257488.1 2 291
NM_012825.4 NP_036957.1 2 283
GenBank mRNA EV243390.1 - 2 350
EV243423.1 - 2 298
L27588.1 - 1 151
U14007.1 - 2 217

View Full Product Details