Gene Transcripts

Gene Symbol
Rassf7
Entrez Gene ID
293623
Gene Name
Ras association (RalGDS/AF-6) domain family (N-terminal) member 7
Gene Aliases
RGD1306244
Chromosome Location
Chr. 1: 214202799 - 214205799 on Build Rnor_6.0
UniGene ID
Rn.24007
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106317.2 NP_001099787.1 1, 2 147
XM_006230537.2 XP_006230599.1 2, 3 247
XM_006230538.2 XP_006230600.1 2, 3 540
XM_006230539.2 XP_006230601.1 1, 2 143
GenBank mRNA BC158743.1 - 1, 2 147

View Full Product Details