Gene Transcripts

Gene Symbol
Lrrc30
Entrez Gene ID
301711
Gene Name
leucine rich repeat containing 30
Gene Aliases
RGD1563832
Chromosome Location
Chr. 9: 115849029 - 115854043 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_237538.4 XP_237538.1 2 608

View Full Product Details