Gene Transcripts

Gene Symbol
Acpt
Entrez Gene ID
308569
Gene Name
acid phosphatase, testicular
Gene Aliases
Chromosome Location
Chr. 1: 100226430 - 100235982 on Build Rnor_6.0
UniGene ID
Rn.154599
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107510.1 NP_001100980.1 6 598
XM_006229051.2 XP_006229113.1 8 946
XM_006229052.2 XP_006229114.1 9 867
XM_006229053.2 XP_006229115.1 8 982
XM_006229054.2 XP_006229116.1 8 1018
XM_008759366.1 XP_008757588.1 7 759
XM_008759367.1 XP_008757589.1 8 982
XM_008759368.1 XP_008757590.1 8 979
XM_008759369.1 XP_008757591.1 7 1811
GenBank mRNA BC168932.1 - 8 1023

View Full Product Details