Gene Transcripts

Gene Symbol
Slc25a45
Entrez Gene ID
689625
Gene Name
solute carrier family 25, member 45
Gene Aliases
Chromosome Location
Chr. 1: 221236741 - 221244564 on Build Rnor_6.0
UniGene ID
Rn.202580
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271179.1 NP_001258108.1 6 717
XM_008760179.1 XP_008758401.1 6 1017
GenBank mRNA FM090288.1 - 2 289

View Full Product Details