Gene Transcripts

Gene Symbol
Sult1d1
Entrez Gene ID
60393
Gene Name
sulfotransferase family 1D, member 1
Gene Aliases
ST1D1, Sult-n, Sultn
Chromosome Location
Chr. 14: 22107418 - 22126011 on Build Rnor_6.0
UniGene ID
Rn.45325
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021769.1 NP_068537.1 7 909
GenBank mRNA U32372.1 - 7 909

View Full Product Details