Gene Transcripts

Gene Symbol
Il9
Entrez Gene ID
116558
Gene Name
interleukin 9
Gene Aliases
Chromosome Location
Chr. 17: 8558827 - 8561950 on Build Rnor_6.0
UniGene ID
Rn.92374
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105747.1 NP_001099217.1 1 120
GenBank mRNA L36460.1 - 1 134

View Full Product Details