Gene Transcripts

Gene Symbol
Map2
Entrez Gene ID
25595
Gene Name
microtubule-associated protein 2
Gene Aliases
MAP2R, Mtap2
Chromosome Location
Chr. 9: 73334908 - 73462972 on Build Rnor_6.0
UniGene ID
Rn.10484
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013066.1 NP_037198.1 5 3238
XM_006245100.2 XP_006245162.1 5 7387
XM_008767197.1 XP_008765419.1 5 3418
XM_008767198.1 XP_008765420.1 5 3418
XM_008767199.1 XP_008765421.1 5 3415
XM_008767200.1 XP_008765422.1 5 3418
XM_008767201.1 XP_008765423.1 5 3415
XM_008767202.1 XP_008765424.1 5 3406
XM_008767203.1 XP_008765425.1 5 3418
XM_008767204.1 XP_008765426.1 4 3172
XM_008767205.1 XP_008765427.1 4 3160
XM_008767206.1 XP_008765428.1 5 3206
GenBank mRNA X51842.1 - 5 3207
X54100.1 - 5 3238

View Full Product Details