Gene Transcripts

Gene Symbol
Higd1b
Entrez Gene ID
287738
Gene Name
HIG1 hypoxia inducible domain family, member 1B
Gene Aliases
RGD1306907
Chromosome Location
Chr. 10: 90929358 - 90931639 on Build Rnor_6.0
UniGene ID
Rn.38891
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105844.1 NP_001099314.1 2, 3 599
XM_008768263.1 XP_008766485.1 3, 4 437
XM_008768264.1 XP_008766486.1 3, 4 322
XM_008768265.1 XP_008766487.1 3, 4 465

View Full Product Details