Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001105966.1 | NP_001099436.1 | 1, 2 | 368 | |
XM_008769625.1 | XP_008767847.1 | 1, 2 | 492 | ||
GenBank mRNA | BC166446.1 | - | 1, 2 | 382 | |
FQ226272.1 | - | 1, 2 | 354 | ||
FQ227495.1 | - | 1, 2 | 372 | ||
FQ230214.1 | - | 1, 2 | 401 | ||
FQ230343.1 | - | 1, 2 | 366 | ||
FQ231443.1 | - | 1, 2 | 355 | ||
FQ232342.1 | - | 1, 2 | 372 | ||
FQ234634.1 | - | 1, 2 | 401 | ||
FQ234923.1 | - | 1, 2 | 365 |