Gene Transcripts

Gene Symbol
Gal
Entrez Gene ID
29141
Gene Name
galanin/GMAP prepropeptide
Gene Aliases
Galn
Chromosome Location
Chr. 1: 218653059 - 218657922 on Build Rnor_6.0
UniGene ID
Rn.8929
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033237.1 NP_150240.1 6 498
GenBank mRNA J03624.1 - 6 498
M18102.1 - 6 478

View Full Product Details