Gene Transcripts

Gene Symbol
Slc10a7
Entrez Gene ID
291942
Gene Name
solute carrier family 10, member 7
Gene Aliases
P7
Chromosome Location
Chr. 19: 32857984 - 33080772 on Build Rnor_6.0
UniGene ID
Rn.23101
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001010948.1 NP_001010948.1 3 287
GenBank mRNA AY825929.1 - 3 287
FQ211514.1 - 479

View Full Product Details