Gene Transcripts

Gene Symbol
Rfx6
Entrez Gene ID
294395
Gene Name
regulatory factor X, 6
Gene Aliases
Rfxdc1
Chromosome Location
Chr. 20: 32709282 - 32764040 on Build Rnor_6.0
UniGene ID
Rn.109159
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106388.1 NP_001099858.1 14 2022
XM_006256444.2 XP_006256506.1 17 2324
XM_006256445.2 XP_006256507.1 10 1575

View Full Product Details