Gene Transcripts

Gene Symbol
Psmd11
Entrez Gene ID
303353
Gene Name
proteasome (prosome, macropain) 26S subunit, non-ATPase, 11
Gene Aliases
Chromosome Location
Chr. 10: 67810655 - 67857562 on Build Rnor_6.0
UniGene ID
Rn.11861
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107027.1 NP_001100497.1 2 414
XM_006246987.2 XP_006247049.1 2 404
XM_006246988.2 XP_006247050.1 2 404

View Full Product Details