Gene Transcripts

Gene Symbol
Gpr126
Entrez Gene ID
308376
Gene Name
G protein-coupled receptor 126
Gene Aliases
Chromosome Location
Chr. 1: 8593342 - 8751534 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006227647.2 XP_006227709.1 19 3159
XM_218313.8 XP_218313.7 20 3244

View Full Product Details