Gene Transcripts

Gene Symbol
Lrrc31
Entrez Gene ID
310256
Gene Name
leucine rich repeat containing 31
Gene Aliases
RGD1561499
Chromosome Location
Chr. 2: 116313489 - 116343076 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006232238.2 XP_006232300.1 5 1642
XM_006232240.2 XP_006232302.1 7 1371
XM_008760950.1 XP_008759172.1 8 3559

View Full Product Details