Gene Transcripts

Gene Symbol
Irf2bpl
Entrez Gene ID
314329
Gene Name
interferon regulatory factor 2 binding protein-like
Gene Aliases
Eap1, Pqcp, RGD1310994
Chromosome Location
Chr. 6: 110901940 - 110904288 on Build Rnor_6.0
UniGene ID
Rn.15682
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012470.1 NP_001012488.1 1 1459
GenBank mRNA BC098759.1 - 1 160

View Full Product Details