Gene Transcripts

Gene Symbol
Hjurp
Entrez Gene ID
316602
Gene Name
Holliday junction recognition protein
Gene Aliases
RGD1305846
Chromosome Location
Chr. 9: 95347739 - 95362165 on Build Rnor_6.0
UniGene ID
Rn.204555
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191813.1 NP_001178742.1 8 1229
XM_006245393.2 XP_006245455.1 8 1395

View Full Product Details