Gene Transcripts

Gene Symbol
Mettl10
Entrez Gene ID
361664
Gene Name
methyltransferase like 10
Gene Aliases
RGD1306300
Chromosome Location
Chr. 1: 204829977 - 204855627 on Build Rnor_6.0
UniGene ID
Rn.21246
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108504.1 NP_001101974.1 6 758
XM_006230441.1 XP_006230503.1 6 753

View Full Product Details