Gene Transcripts

Gene Symbol
Smg9
Entrez Gene ID
365215
Gene Name
SMG9 nonsense mediated mRNA decay factor
Gene Aliases
RGD1309888
Chromosome Location
Chr. 1: 81259447 - 81282893 on Build Rnor_6.0
UniGene ID
Rn.32677
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014243.1 NP_001014265.1 7 842
XM_006228450.2 XP_006228512.1 7 1773
GenBank mRNA BC087051.1 - 7 842

View Full Product Details