Gene Transcripts

Gene Symbol
Dear
Entrez Gene ID
446170
Gene Name
dual endothelin 1, angiotensin II receptor
Gene Aliases
Chromosome Location
Chr. 2: 184243976 - 184247228 on Build Rnor_6.0
UniGene ID
Rn.215682
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004448.1 NP_001004448.1 1 2726
GenBank mRNA AY664492.1 - 1 2726

View Full Product Details