Gene Transcripts

Gene Symbol
Vom1r30
Entrez Gene ID
494230
Gene Name
vomeronasal 1 receptor 30
Gene Aliases
V1rl1
Chromosome Location
Chr. 1: 63333283 - 63334176 on Build Rnor_6.0
UniGene ID
Rn.129435
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009511.1 NP_001009511.1 1 553
GenBank mRNA AY510321.1 - 1 553

View Full Product Details