Gene Transcripts

Gene Symbol
Nsg2
Entrez Gene ID
497878
Gene Name
neuron specific gene family member 2
Gene Aliases
Chromosome Location
Chr. 10: 15867885 - 15928169 on Build Rnor_6.0
UniGene ID
Rn.14657
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034152.1 NP_001029324.1 3 336
GenBank mRNA BC105916.1 - 3 336

View Full Product Details