Gene Transcripts

Gene Symbol
Arhgap30
Entrez Gene ID
498282
Gene Name
Rho GTPase activating protein 30
Gene Aliases
RGD1561419
Chromosome Location
Chr. 13: 89774764 - 89796113 on Build Rnor_6.0
UniGene ID
Rn.131539
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109077.1 NP_001102547.1 10 1361
GenBank mRNA FQ211641.1 - 1369

View Full Product Details