Gene Transcripts

Gene Symbol
Selm
Entrez Gene ID
498398
Gene Name
selenoprotein M
Gene Aliases
RGD1565037
Chromosome Location
Chr. 14: 83752393 - 83754975 on Build Rnor_6.0
UniGene ID
Rn.8166
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001115013.1 NP_001108485.1 2 194
GenBank mRNA BI282694.1 - 4 499
CB607044.1 - 2 166
FQ223498.1 - 2 149

View Full Product Details