Gene Transcripts

Gene Symbol
Wisp3
Entrez Gene ID
499461
Gene Name
WNT1 inducible signaling pathway protein 3
Gene Aliases
RGD1564120
Chromosome Location
Chr. 20: 44237491 - 44255064 on Build Rnor_6.0
UniGene ID
Rn.141375
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170483.1 NP_001163954.1 4 698

View Full Product Details