Gene Transcripts

Gene Symbol
Tas2r138
Entrez Gene ID
500091
Gene Name
taste receptor, type 2, member 138
Gene Aliases
T2R38, Tas2r38
Chromosome Location
Chr. 4: 68831221 - 68832216 on Build Rnor_6.0
UniGene ID
Rn.156466
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024685.1 NP_001019856.1 1 349
GenBank mRNA AY916512.1 - 1 349

View Full Product Details