Gene Transcripts

Gene Symbol
Kdm1a
Entrez Gene ID
500569
Gene Name
lysine (K)-specific demethylase 1A
Gene Aliases
Aof2, Kdm1, RGD1562975
Chromosome Location
Chr. 5: 154909003 - 154965171 on Build Rnor_6.0
UniGene ID
Rn.203461
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130098.1 NP_001123570.1 9 1239
GenBank mRNA EF688598.1 - 9 1239

View Full Product Details