Gene Transcripts

Gene Symbol
Rwdd4
Entrez Gene ID
502084
Gene Name
RWD domain containing 4A
Gene Aliases
Rwdd4a
Chromosome Location
Chr. 16: 47856522 - 47874856 on Build Rnor_6.0
UniGene ID
Rn.15503
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034994.1 NP_001030166.1 5 600
GenBank mRNA BC092581.1 - 5 600
FQ229862.1 - 5 641

View Full Product Details