Gene Transcripts

Gene Symbol
Vom2r51
Entrez Gene ID
502891
Gene Name
vomeronasal 2 receptor, 51
Gene Aliases
RGD1561712
Chromosome Location
Chr. 4: 156515299 - 156563438 on Build Rnor_6.0
UniGene ID
Rn.217944
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099510.1 NP_001092980.1 1 196

View Full Product Details