Gene Transcripts

Gene Symbol
Sage1
Entrez Gene ID
681682
Gene Name
sarcoma antigen 1
Gene Aliases
Chromosome Location
Chr. X: 158923560 - 158931082 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008773664.1 XP_008771886.1 2 307

View Full Product Details