Gene Transcripts

Gene Symbol
Tcap
Entrez Gene ID
688173
Gene Name
titin-cap
Gene Aliases
Chromosome Location
Chr. 10: 86337705 - 86338414 on Build Rnor_6.0
UniGene ID
Rn.216482
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271277.1 NP_001258206.1 2 265
GenBank mRNA FQ215695.1 - 2 263
FQ216780.1 - 2 265

View Full Product Details