Assay Design



Gene Transcripts

Gene Symbol
GAB2
Entrez Gene ID
700197
Gene Name
GRB2 associated binding protein 2
Gene Aliases
-
Chromosome Location
Chr.14: 71295542 - 71498047 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015115335.2 XP_014970821.1 - 716 59
GenBank mRNA JU497118.1 - - 645 59
JU497119.1 - - 645 59

View Full Product Details