Assay Design



Gene Transcripts

Gene Symbol
LGI2
Entrez Gene ID
693308
Gene Name
leucine rich repeat LGI family member 2
Gene Aliases
-
Chromosome Location
Chr.5: 23947632 - 23980632 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001082016.4 XP_001082016.1 - 448 62
GenBank mRNA JU327322.1 - - 180 62
JV718028.1 - - 157 62

View Full Product Details