Assay Design



Gene Transcripts

Gene Symbol
MBLAC2
Entrez Gene ID
695437
Gene Name
metallo-beta-lactamase domain containing 2
Gene Aliases
EGK_16669
Chromosome Location
Chr.6: 87325148 - 87344279 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193835.2 NP_001180764.1 - 932 59
GenBank mRNA JU321955.1 - - 670 59
JU470921.1 - - 488 59

View Full Product Details