Assay Design
Gene Transcripts
- Gene Symbol
- LRRC14
- Entrez Gene ID
-
703587
- Gene Name
- leucine rich repeat containing 14
- Gene Aliases
-
EGK_19383
- Chromosome Location
-
Chr.8: 144832482 - 144839662 on Build Mmul_10
- Gene Symbol
- LRRC24
- Entrez Gene ID
-
703465
- Gene Name
- leucine rich repeat containing 24
- Gene Aliases
-
-
- Chromosome Location
-
Chr.8: 144836539 - 144841891 on Build Mmul_10
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
JV599337.1 |
- |
- |
1034 |
91 |
View Full Product Details