Assay Design



Gene Transcripts

Gene Symbol
MB21D2
Entrez Gene ID
706284
Gene Name
Mab-21 domain containing 2
Gene Aliases
C3orf59, EGK_12225
Chromosome Location
Chr.2: 5040639 - 5160693 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194011.2 NP_001180940.2 - 537 72

View Full Product Details