Assay Design



Gene Transcripts

Gene Symbol
KMT2D
Entrez Gene ID
710795
Gene Name
lysine methyltransferase 2D
Gene Aliases
MLL2
Chromosome Location
Chr.11: 48628136 - 48670741 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015151647.2 XP_015007133.2 - 14845 79
GenBank mRNA JU494186.1 - - 1581 79
JV072336.1 - - 1642 79

View Full Product Details