Assay Design



Gene Transcripts

Gene Symbol
TESK1
Entrez Gene ID
697206
Gene Name
testis associated actin remodelling kinase 1
Gene Aliases
-
Chromosome Location
Chr.15: 48593379 - 48598134 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001265761.1 NP_001252690.1 - 483 61
XM_015117343.2 XP_014972829.1 - 540 61
GenBank mRNA JV565173.1 - - 483 61

View Full Product Details