Assay Design



Gene Transcripts

Gene Symbol
GUCA2B
Entrez Gene ID
699525
Gene Name
guanylate cyclase activator 2B
Gene Aliases
EGK_00632
Chromosome Location
Chr.1: 182394186 - 182396560 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001087987.3 XP_001087987.1 - 129 73

View Full Product Details