Assay Design



Gene Transcripts

Gene Symbol
ELAVL2
Entrez Gene ID
708195
Gene Name
ELAV like RNA binding protein 2
Gene Aliases
-
Chromosome Location
Chr.15: 59867378 - 60026998 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015117600.2 XP_014973086.1 - 1405 103
XM_015117607.2 XP_014973093.1 - 1314 103
XM_028834316.1 XP_028690149.1 - 1112 103
XM_028834317.1 XP_028690150.1 - 1402 103
XM_028834324.1 XP_028690157.1 - 968 103
XM_028834325.1 XP_028690158.1 - 1021 103
GenBank mRNA JU320297.1 - - 992 103
JV497440.1 - - 1038 103

View Full Product Details