Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001194040.1 | NP_001180969.1 | - | 434 | 66 |
XM_001095958.4 | XP_001095958.1 | - | 391 | 66 | |
XM_028843211.1 | XP_028699044.1 | - | 522 | 66 | |
XM_028843212.1 | XP_028699045.1 | - | 344 | 66 | |
GenBank mRNA | JU335538.1 | - | - | 261 | 66 |
JU476466.1 | - | - | 264 | 66 | |
JV513579.1 | - | - | 218 | 66 | |
JV587837.1 | - | - | 260 | 66 | |
JV656419.1 | - | - | 165 | 66 |