Assay Design



Gene Transcripts

Gene Symbol
STOML2
Entrez Gene ID
698458
Gene Name
stomatin like 2
Gene Aliases
EGK_07581
Chromosome Location
Chr.15: 49127911 - 49131166 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260882.2 NP_001247811.1 - 631 66
GenBank mRNA JU321476.1 - - 610 66
JU472282.1 - - 627 66
JV043525.1 - - 634 66
JV669581.1 - - 637 66

View Full Product Details