Assay Design



Gene Transcripts

Gene Symbol
STMN2
Entrez Gene ID
699264
Gene Name
stathmin 2
Gene Aliases
EGK_19065
Chromosome Location
Chr.8: 80065521 - 80120068 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001265775.1 NP_001252704.1 - 75 66
GenBank mRNA JV569723.1 - - 75 66
JV667332.1 - - 75 66

View Full Product Details