Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_001109452.4 | XP_001109452.2 | - | 1409 | 84 |
XM_001109551.4 | XP_001109551.2 | - | 1044 | 84 | |
XM_015113046.2 | XP_014968532.1 | - | 1412 | 84 | |
XM_015113047.2 | XP_014968533.1 | - | 2615 | 84 | |
XM_015113048.2 | XP_014968534.1 | - | 1502 | 84 | |
GenBank mRNA | JU319712.1 | - | - | 1129 | 84 |
JU471147.1 | - | - | 1211 | 84 | |
JV043819.1 | - | - | 1161 | 84 | |
JV529552.1 | - | - | 1212 | 84 | |
JV668167.1 | - | - | 1220 | 84 |