Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_001092818.4 | XP_001092818.1 | - | 661 | 84 |
GenBank mRNA | JU332625.1 | - | - | 628 | 84 |
JU474084.1 | - | - | 602 | 84 | |
JV045994.1 | - | - | 613 | 84 | |
JV604088.1 | - | - | 631 | 84 |