Assay Design



Gene Transcripts

Gene Symbol
UBE2B
Entrez Gene ID
710678
Gene Name
ubiquitin conjugating enzyme E2 B
Gene Aliases
EGK_16859
Chromosome Location
Chr.6: 131834170 - 131855690 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194433.3 NP_001181362.1 - 554 124
GenBank mRNA JU320381.1 - - 183 124
JU471302.1 - - 242 124

View Full Product Details