Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198667.1 | NP_001185596.1 | - | 454 | 73 |
GenBank mRNA | EG703550.1 | - | - | 454 | 73 |
JU477632.1 | - | - | 427 | 73 | |
JV048565.1 | - | - | 411 | 73 | |
JV565304.1 | - | - | 436 | 73 |