Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001261540.1 | NP_001248469.1 | - | 297 | 60 |
XM_015141466.2 | XP_014996952.2 | - | 476 | 60 | |
GenBank mRNA | JU319705.1 | - | - | 297 | 60 |
JV567215.1 | - | - | 379 | 60 |